siRNA Details
| Gene Symbol: Mzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290452.1 | NP_001277381.1 | 4, 5 | 924 | ||
| NM_145819.2 | NP_665818.2 | 5, 6 | 1108 | |||
| GenBank mRNA | AB007407.1 | - | 4, 5 | 1327 | ||
| AB088478.1 | - | 5, 6 | 1009 | |||
| AK145014.1 | - | 5, 6 | 989 | |||
| BC129904.1 | - | 4, 5 | 674 | 40111654 | ||
| BC129905.1 | - | 5, 6 | 858 | 40111655 | ||

Mzf1
Genomic Map