siRNA Details
| Gene Symbol: Mzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290453.1 | NP_001277382.1 | 3 | 799 | ||
| NM_145819.2 | NP_665818.2 | 3 | 799 | |||
| GenBank mRNA | AB007407.1 | - | 2 | 1018 | ||
| AB088478.1 | - | 3 | 700 | |||
| AK030410.1 | - | 3 | 800 | |||
| AK145014.1 | - | 3 | 680 | |||
| AK158837.1 | - | 1 | 2051 | |||
| BC060649.1 | - | 3 | 759 | |||
| BC129905.1 | - | 3 | 549 | 40111655 | ||

Mzf1
Genomic Map