siRNA Details
| Gene Symbol: Asl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133768.4 | NP_598529.1 | 4 | 322 | ||
| GenBank mRNA | AK010922.1 | - | 4 | 336 | ||
| AK076453.1 | - | 4 | 337 | |||
| AK138423.1 | - | 4 | 319 | |||
| AK140160.1 | - | 4 | 337 | |||
| AK148764.1 | - | 4 | 306 | |||
| AK168739.1 | - | 4 | 317 | |||
| AK186886.1 | - | 2 | 173 | |||
| BC016670.1 | - | 4 | 322 | 2648753 | ||

Asl
Genomic Map