siRNA Details
| Gene Symbol: Art4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026639.2 | NP_080915.1 | 2 | 497 | ||
| GenBank mRNA | AK004744.1 | - | 2 | 451 | ||
| AK019467.1 | - | 2 | 498 | |||
| BC046306.1 | - | 2 | 418 | 6306234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Art4| Gene Symbol: Art4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026639.2 | NP_080915.1 | 2 | 497 | ||
| GenBank mRNA | AK004744.1 | - | 2 | 451 | ||
| AK019467.1 | - | 2 | 498 | |||
| BC046306.1 | - | 2 | 418 | 6306234 | ||