siRNA Details
| Gene Symbol: Art3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181728.2 | NP_859417.2 | 7 | 1073 | ||
| GenBank mRNA | AJ311769.1 | - | 5 | 886 | ||
| AJ311770.1 | - | 5 | 886 | |||
| AJ311771.1 | - | 5 | 886 | |||
| AJ311773.1 | - | 5 | 886 | |||
| AJ311774.1 | - | 4 | 177 | |||
| AK016257.1 | - | 7 | 1066 | |||
| BC141069.1 | - | 5 | 933 | 9055681 | ||
| BC145327.1 | - | 5 | 933 | 9053883 | ||
| BE625316.1 | - | 4 | 198 | |||

Art3
Genomic Map