siRNA Details
| Gene Symbol: Dnah5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133365.3 | NP_579943.3 | 24 | 3962 | ||
| GenBank mRNA | AF466704.1 | - | 24 | 3963 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnah5| Gene Symbol: Dnah5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133365.3 | NP_579943.3 | 24 | 3962 | ||
| GenBank mRNA | AF466704.1 | - | 24 | 3963 | ||