siRNA Details
| Gene Symbol: Raf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029780.3 | NP_084056.1 | 12 | 1733 | ||
| GenBank mRNA | AB057655.1 | - | 11 | 1278 | ||
| AB057663.2 | - | 12 | 1503 | |||
| AK018272.1 | - | 3 | 289 | |||
| AK141745.1 | - | 12 | 1734 | |||
| AK148327.1 | - | 642 | ||||
| BC015273.1 | - | 12 | 1612 | 4222659 | ||
| BC056439.1 | - | 12 | 1650 | |||
| BC092040.1 | - | 12 | 1598 | 2645313 | ||

Raf1
Genomic Map