siRNA Details
| Gene Symbol: Gpr18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182806.1 | NP_877958.1 | 2 | 307 | ||
| GenBank mRNA | AK156039.1 | - | 2 | 380 | ||
| AK156868.1 | - | 2 | 372 | |||
| AK157029.1 | - | 2 | 315 | |||
| AK172401.1 | - | 2 | 310 | |||
| BC034872.1 | - | 2 | 307 | 1397739 | ||

Gpr18
Genomic Map