siRNA Details
| Gene Symbol: Slc35b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016752.1 | NP_058032.1 | 7, 8 | 853 | ||
| GenBank mRNA | AK002961.1 | - | 7, 8 | 847 | ||
| AK007711.1 | - | 7, 8 | 859 | |||
| AK160124.1 | - | 7, 8 | 920 | |||
| BC002029.1 | - | 7, 8 | 834 | 3592141 | ||
| D87990.1 | - | 7, 8 | 853 | |||

Slc35b1
Genomic Map