siRNA Details
| Gene Symbol: P2RY11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002566.4 | NP_002557.2 | 2 | 1582 | ||
| GenBank mRNA | BC073827.1 | - | 2 | 1565 | 5176598 | |
| Gene Symbol: PPAN-P2RY11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040664.2 | NP_001035754.1 | 13 | 2813 | ||
| NM_001198690.1 | NP_001185619.1 | 13 | 2875 | |||
More Information
Set Membership: |
|

P2RY11
Genomic Map