siRNA Details
| Gene Symbol: Pp2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173449.3 | NP_775625.1 | 2 | 747 | ||
| GenBank mRNA | AK076579.1 | - | 2 | 748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pp2d1| Gene Symbol: Pp2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173449.3 | NP_775625.1 | 2 | 747 | ||
| GenBank mRNA | AK076579.1 | - | 2 | 748 | ||