siRNA Details
| Gene Symbol: Adrbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290818.1 | NP_001277747.1 | 9 | 872 | ||
| NM_130863.2 | NP_570933.1 | 10 | 944 | |||
| GenBank mRNA | AF333028.1 | - | 9 | 664 | ||
| AK155691.1 | - | 9 | 873 | |||
| AK193363.1 | - | 4 | 185 | |||
| BC003196.1 | - | 9 | 660 | |||
| BC033272.1 | - | 8 | 988 | |||
| BC053922.1 | - | 10 | 677 | 6309555 | ||

Adrbk1
Genomic Map