siRNA Details
| Gene Symbol: Adrbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290818.1 | NP_001277747.1 | 16 | 1592 | ||
| NM_130863.2 | NP_570933.1 | 17 | 1664 | |||
| GenBank mRNA | AF333028.1 | - | 16 | 1384 | ||
| AK155691.1 | - | 16 | 1593 | |||
| BC003196.1 | - | 16 | 1380 | |||
| BC033272.1 | - | 15 | 1708 | |||
| BC053922.1 | - | 17 | 1397 | 6309555 | ||

Adrbk1
Genomic Map