siRNA Details
| Gene Symbol: Shroom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290684.1 | NP_001277613.1 | 4 | 2770 | ||
| NM_172441.3 | NP_766029.2 | 4 | 2770 | |||
| GenBank mRNA | CK635066.1 | - | 1 | 35 | ||
| DQ358971.1 | - | 4 | 2751 | |||
| EF071946.1 | - | 4 | 2770 | |||

Shroom2
Genomic Map