siRNA Details
| Gene Symbol: Acat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144784.3 | NP_659033.1 | 8 | 845 | ||
| GenBank mRNA | AJ510150.1 | - | 8 | 837 | ||
| AK032097.1 | - | 8 | 816 | |||
| AK075593.1 | - | 8 | 818 | |||
| AK081715.1 | - | 8 | 845 | |||
| AK146241.1 | - | 8 | 800 | |||
| AK163402.1 | - | 8 | 849 | |||
| AK169771.1 | - | 8 | 838 | |||
| BC024429.1 | - | 8 | 848 | |||
| BC024763.1 | - | 8 | 817 | 5365469 | ||

Acat1
Genomic Map