siRNA Details
Gene Symbol: Olfr153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206823.1 | NP_996559.1 | 1 | 268 | ||
GenBank mRNA | BC147769.1 | - | 1 | 507 | 9056708 | |
BC147777.1 | - | 1 | 507 | 9056711 |
Gene Symbol: Olfr1131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146658.2 | NP_666869.2 | 1 | 268 |
