siRNA Details
| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175481.5 | NP_780690.2 | 13 | 1522 | ||
| GenBank mRNA | AK032029.1 | - | 13 | 1523 | ||
| AK080582.1 | - | 13 | 1651 | |||
| BC118010.2 | - | 12 | 1437 | 40096690 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grik4| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175481.5 | NP_780690.2 | 13 | 1522 | ||
| GenBank mRNA | AK032029.1 | - | 13 | 1523 | ||
| AK080582.1 | - | 13 | 1651 | |||
| BC118010.2 | - | 12 | 1437 | 40096690 | ||