siRNA Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127338.1 | NP_001120810.1 | 14 | 1293 | ||
| NM_138600.4 | NP_613066.2 | 12 | 1099 | |||
| GenBank mRNA | AK004991.1 | - | 14 | 1227 | ||
| AK145873.1 | - | 12 | 1042 | |||
| AK160117.1 | - | 12 | 1026 | |||
| AK169195.1 | - | 12 | 1042 | |||
| AK183099.1 | - | 2 | 100 | |||
| BC012407.1 | - | 12 | 1061 | 4191228 | ||

Aldh7a1
Genomic Map