siRNA Details
| Gene Symbol: H2-M10.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201608.2 | NP_963902.1 | 2 | 262 | ||
| GenBank mRNA | AF539599.1 | - | 2 | 256 | ||
| AY211080.1 | - | 2 | 261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2-M10.3| Gene Symbol: H2-M10.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201608.2 | NP_963902.1 | 2 | 262 | ||
| GenBank mRNA | AF539599.1 | - | 2 | 256 | ||
| AY211080.1 | - | 2 | 261 | |||