siRNA Details
Gene Symbol: H2-M10.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201608.2 | NP_963902.1 | 3 | 527 | ||
GenBank mRNA | AF539599.1 | - | 3 | 521 | ||
AY211080.1 | - | 3 | 526 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: H2-M10.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201608.2 | NP_963902.1 | 3 | 527 | ||
GenBank mRNA | AF539599.1 | - | 3 | 521 | ||
AY211080.1 | - | 3 | 526 |