siRNA Details
| Gene Symbol: Adam33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163529.1 | NP_001157001.1 | 18 | 2415 | ||
| NM_033615.2 | NP_291093.2 | 19 | 2493 | |||
| GenBank mRNA | AB059632.1 | - | 19 | 2493 | ||
| AB059633.1 | - | 18 | 2415 | |||
| AF386072.1 | - | 15 | 1857 | |||
| AF472524.1 | - | 19 | 2225 | |||
| AK048367.1 | - | 14 | 1849 | |||
| AK053825.1 | - | 15 | 2912 | |||
| AY382193.1 | - | 18 | 2146 | |||
| AY382194.1 | - | 19 | 2224 | |||
| BC138139.1 | - | 18 | 2138 | 8861158 | ||

Adam33
Genomic Map