siRNA Details
| Gene Symbol: Adgrv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054053.4 | NP_473394.3 | 8, 9 | 1631 | ||
| GenBank mRNA | AB086166.2 | - | 8, 9 | 1626 | ||
| AB086167.1 | - | 8, 9 | 1626 | |||
| AF405692.1 | - | 4, 5 | 1776 | |||
| AF435926.1 | - | 8, 9 | 1632 | |||

Adgrv1
Genomic Map