siRNA Details
| Gene Symbol: Adgrv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054053.4 | NP_473394.3 | 14 | 2763 | ||
| GenBank mRNA | AB086167.1 | - | 14 | 2758 | ||
| AF405692.1 | - | 10 | 2908 | |||
| AF405693.1 | - | 4 | 562 | |||
| AF435926.1 | - | 14 | 2764 | |||

Adgrv1
Genomic Map