siRNA Details
| Gene Symbol: Srsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001078167.2 | NP_001071635.1 | 3 | 991 | ||
| NM_173374.4 | NP_775550.2 | 3 | 991 | |||
| GenBank mRNA | AK018176.1 | - | 3 | 660 | ||
| AK053674.1 | - | 3 | 682 | |||
| AK078715.1 | - | 3 | 648 | |||
| AK150535.1 | - | 3 | 694 | |||
| AK169203.1 | - | 3 | 648 | |||
| AK169623.1 | - | 3 | 649 | |||
| BC046773.1 | - | 3 | 623 | 5708288 | ||
| BC058627.1 | - | 3 | 657 | 6401674 | ||
| X66091.1 | - | 3 | 604 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109552, XM_008768114, FM055064.

Srsf1
Genomic Map