siRNA Details
| Gene Symbol: Pcca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144844.2 | NP_659093.2 | 18 | 1592 | ||
| GenBank mRNA | AK148002.1 | - | 18 | 1582 | ||
| AK183812.1 | - | 1 | 19 | |||
| AK215585.1 | - | 1 | 19 | |||
| AY046947.1 | - | 18 | 1609 | |||
| BC006915.1 | - | 18 | 1589 | 3601148 | ||
| BC049802.1 | - | 18 | 1558 | 6332730 | ||

Pcca
Genomic Map