siRNA Details
| Gene Symbol: Pcca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144844.2 | NP_659093.2 | 10 | 827 | ||
| GenBank mRNA | AK148002.1 | - | 10 | 817 | ||
| AY046947.1 | - | 10 | 844 | |||
| BC006915.1 | - | 10 | 824 | 3601148 | ||
| BC049802.1 | - | 10 | 793 | 6332730 | ||

Pcca
Genomic Map