siRNA Details
| Gene Symbol: Etfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026695.3 | NP_080971.2 | 5 | 566 | ||
| GenBank mRNA | AA207939.1 | - | 5 | 449 | ||
| AK002407.1 | - | 5 | 508 | |||
| AK013284.1 | - | 5 | 530 | |||
| AK150293.1 | - | 5 | 509 | |||
| AK192361.1 | - | 1 | 57 | |||
| BC049237.1 | - | 5 | 495 | 4241552 | ||
| BC069877.1 | - | 5 | 525 | 5370580 | ||

Etfb
Genomic Map