siRNA Details
| Gene Symbol: Etfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026695.3 | NP_080971.2 | 6 | 746 | ||
| GenBank mRNA | AK002407.1 | - | 6 | 688 | ||
| AK013284.1 | - | 6 | 710 | |||
| AK150293.1 | - | 6 | 689 | |||
| BC049237.1 | - | 6 | 675 | 4241552 | ||
| BC069877.1 | - | 6 | 705 | 5370580 | ||

Etfb
Genomic Map