siRNA Details
| Gene Symbol: Lims1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193303.1 | NP_001180232.1 | 2 | 379 | ||
| NM_026148.3 | NP_080424.2 | 2 | 373 | |||
| NM_201242.1 | NP_957694.1 | 2 | 397 | |||
| GenBank mRNA | AK009390.1 | - | 2 | 380 | ||
| AK014954.1 | - | 2 | 374 | |||
| AK082932.1 | - | 2 | 242 | |||
| AK088939.1 | - | 2 | 248 | |||
| AK151235.1 | - | 2 | 248 | |||
| AK151938.1 | - | 2 | 240 | |||
| AK152679.1 | - | 2 | 240 | |||
| AK153346.1 | - | 2 | 248 | |||
| BC005621.1 | - | 2 | 228 | 3711611 | ||
| BC067393.1 | - | 2 | 171 | |||
| DQ003608.1 | - | 2 | 364 | |||
| EU122953.1 | - | 2 | 363 | |||
| EU122954.1 | - | 2 | 231 | |||

Lims1
Genomic Map