siRNA Details
| Gene Symbol: Chrna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176844.4 | NP_789814.2 | 5 | 726 | ||
| GenBank mRNA | AF204689.1 | - | 4 | 491 | ||
| AK080900.1 | - | 2 | 435 | |||
| AY574264.1 | - | 5 | 509 | |||
| BC106839.2 | - | 5 | 509 | 40045716 | ||
| BC106853.2 | - | 5 | 509 | 40046382 | ||
| DQ318788.1 | - | 5 | 679 | |||

Chrna5
Genomic Map