siRNA Details
| Gene Symbol: Kcnq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152923.2 | NP_690887.2 | 1 | 329 | ||
| GenBank mRNA | AY118171.1 | - | 1 | 329 | ||
| BC116306.1 | - | 1 | 320 | 40095195 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnq3| Gene Symbol: Kcnq3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152923.2 | NP_690887.2 | 1 | 329 | ||
| GenBank mRNA | AY118171.1 | - | 1 | 329 | ||
| BC116306.1 | - | 1 | 320 | 40095195 | ||