siRNA Details
| Gene Symbol: Slc18a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153054.2 | NP_694694.1 | 13 | 1579 | ||
| GenBank mRNA | AK076433.1 | - | 12 | 1304 | ||
| AY779336.1 | - | 11 | 1237 | |||
| BC027192.1 | - | 13 | 1517 | 4239930 | ||
| BC031146.1 | - | 13 | 1547 | 4974776 | ||

Slc18a1
Genomic Map