siRNA Details
| Gene Symbol: Gabra5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176942.4 | NP_795916.1 | 10 | 1617 | ||
| GenBank mRNA | AK038476.1 | - | 10 | 1619 | ||
| AK083185.1 | - | 10 | 1404 | |||
| BC062112.1 | - | 10 | 1615 | 6815819 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabra5| Gene Symbol: Gabra5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176942.4 | NP_795916.1 | 10 | 1617 | ||
| GenBank mRNA | AK038476.1 | - | 10 | 1619 | ||
| AK083185.1 | - | 10 | 1404 | |||
| BC062112.1 | - | 10 | 1615 | 6815819 | ||