siRNA Details
| Gene Symbol: Slc9a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177084.3 | NP_796058.1 | 8 | 1885 | ||
| GenBank mRNA | AK085681.1 | - | 8 | 1911 | ||
| AK141534.1 | - | 8 | 1886 | |||
| AK170522.1 | - | 6 | 724 | |||
| BC120543.1 | - | 8 | 1803 | 40129466 | ||

Slc9a4
Genomic Map