siRNA Details
| Gene Symbol: Hspa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030201.3 | NP_084477.1 | 3 | 596 | ||
| GenBank mRNA | AK021006.1 | - | 3 | 568 | ||
| AK030553.1 | - | 3 | 569 | |||
| AK034643.1 | - | 3 | 576 | |||
| AK049011.1 | - | 3 | 564 | |||
| AK167839.1 | - | 3 | 539 | |||
| BC039701.1 | - | 2 | 537 | |||
| BC085181.1 | - | 3 | 556 | 30646642 | ||
| BC094508.1 | - | 3 | 539 | |||

Hspa13
Genomic Map