siRNA Details
| Gene Symbol: Atp6v1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134157.2 | NP_598918.1 | 13, 14 | 1443 | ||
| GenBank mRNA | AF435091.1 | - | 13, 14 | 1443 | ||
| AK052604.1 | - | 13, 14 | 1414 | |||
| AK052707.1 | - | 13, 14 | 1405 | |||
| AK078810.1 | - | 13, 14 | 1471 | |||
| AK085549.1 | - | 13, 14 | 1444 | |||
| AK168980.1 | - | 13, 14 | 1471 | |||
| BC004789.1 | - | 10, 11 | 1037 | |||
| BC017127.1 | - | 13, 14 | 1428 | 4489144 | ||
| BC062202.1 | - | 13, 14 | 1402 | 2650839 | ||

Atp6v1b1
Genomic Map