siRNA Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139145.4 | NP_631884.1 | 9 | 2116 | ||
| GenBank mRNA | AB066227.1 | - | 8 | 1976 | ||
| AK052424.1 | - | 10 | 2412 | |||
| AK158212.1 | - | 10 | 2233 | |||
| AK168925.1 | - | 6 | 1011 | |||
| BC050090.1 | - | 9 | 2117 | 30012868 | ||

Hlcs
Genomic Map