siRNA Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139145.4 | NP_631884.1 | 8 | 1935 | ||
| GenBank mRNA | AB066227.1 | - | 7 | 1795 | ||
| AK052424.1 | - | 9 | 2231 | |||
| AK158212.1 | - | 9 | 2052 | |||
| AK168925.1 | - | 5 | 830 | |||
| BC050090.1 | - | 8 | 1936 | 30012868 | ||

Hlcs
Genomic Map