Learn More
Medchemexpress LLC Racemic abscisic acid | 14375-45-2 | MFCD00075619 | 99.9% | 264.32 g/mol | C15H20O4 | 10 MG
Supplier: Medchemexpress LLC HYN254910MG
(±)-Abscisic acid is the racemic form of the plant hormone abscisic acid supplied as a high-purity research chemical in a 10 mg quantity. It is used in biochemical and pharmacological studies to investigate stress signaling, metabolism, and glycemic regulation. The material is provided as a powder with defined solubility and storage recommendations for in vitro and in vivo use.
- High purity (99.91%) suitable for research applications.
- Small-scale 10 mg quantity for screening and pilot studies.
- Powder form for flexible reconstitution in common solvents.
- Soluble in DMSO (≈250 mg/mL) for in vitro assays.
- Documented storage conditions to maintain stability.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.