Learn More
Medchemexpress LLC Beta-D-glucan | 9041-22-9 | MFCD00466918 | 99.0% | (C6H10O5)n | 10MG
SDP
Supplier:  Medchemexpress LLC HY13941310MG
Encompass_Preferred
β-D-Glucan is a non-digestible polysaccharide (CAS 9041-22-9) provided as a research-grade reagent. It is supplied as a solid, white to light-yellow powder with reported purity of 99.0% and is commonly used in immunology research and as an enteric delivery vehicle. Store protected from light at 4°C; in solvent, follow recommended cold-storage conditions.
Catalog No. 50-002-60572