Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Tyrphostin B44 | 133550-37-5 | MFCD00209855 | 97.0% | 308.33 | C18H16N2O3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY101007A10MG

Encompass_Preferred

(+)-Tyrphostin B44 is a small-molecule epidermal growth factor receptor (EGFR) inhibitor used in laboratory research to probe EGFR signaling and assess antitumor activity. It is supplied as a solid or as a DMSO solution and is intended for research use only.

  • EGFR inhibitor for preclinical research.
  • Used to study EGFR signaling and antitumor activity.
  • Available as a solid or 10 mM solution in DMSO.
  • Reported purity 97.0%.
  • Molecular weight 308.33 g/mol.
  • CAS number 133550-37-5.
  • Soluble in DMSO; handle according to standard laboratory safety practices.

Catalog No. 50-002-12953


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.