Learn More
Teknova 0.01M PBS with 0.02% Tween-80. 150mL in 250mL Bottle, Sterile 8 pack.
SDP
Supplier:  Teknova P38738PK
Encompass_Preferred
0.850% Sodium Chloride, 0.120% Sodium Phosphate Dibasic, 0.022% Sodium Phosphate Monobasic, 0.020% Tween-80
Catalog No. 50-238-7242
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.