Learn More
Molecular Dimensions 0.04MM MOUNTED LITHO LOOPS
SDP
Supplier:  Molecular Dimensions MD7131
Encompass_Preferred
Protein Crystallography. 0.04mm LithoLoops mounted on 18mm stainless steel microtubes. Mounted in 18mm SPINE microtubes (pins). For use with Cryocaps. Just superglue the pin into a cyrocap and you're ready to go.
Catalog No. NC0581707