Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

0.07% n-Hexylamine in Chloroform, Reagents

Catalog No. 01334694
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-694 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-694 Supplier Reagents Holdings Llc Supplier No. CN0007004C
Only null left
Add to Cart
Add to Cart

0.07% n-Hexylamine in Chloroform, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-66-3 , 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym n-Hexylamine, 1-Aminohexane
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Color Colorless
Boiling Point 61°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 1.484
Grade Reagent
Physical Form Liquid
Chemical Name or Material 0.07% n-Hexylamine in Chloroform
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.