Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

0.07% n-Hexylamine in HPLC Grade Acetonitrile, Reagents

Catalog No. 01334697
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-697 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-697 Supplier Reagents Holdings Llc Supplier No. CN0004004C
Only null left
Add to Cart
Add to Cart

0.07% n-Hexylamine in HPLC Grade Acetonitrile, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-05-8 , 111-26-2
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym n-Hexylamine, 1-Aminohexane
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Color Colorless
Boiling Point 82°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.7858
Grade Reagent
Physical Form Liquid
Chemical Name or Material 0.07% n-Hexylamine in HPLC Grade Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.