Learn More
Thermo Scientific™ 0.1 mL Individual UTW Tubes
Catalog No. TUC0011 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
White
Catalog No. Color
TUC0011 White
1 options
Catalog No. TUC0011 Supplier Thermo Scientific™ Supplier No. TUC0011
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Ultra thin wall individual PCR tubes with attached flat caps.
UTW vessels have walls that are 0.15 mm thin where it matters the most - at the contact point between the thermal cycler block and the reactants. Thinner walls offer less thermal resistance for transferring heat from the block to the sample within the tube. This results in the ability to perform very rapid protocols by reducing incubation times during PCR.
HIghlights
  • Caps form a secure seal, yet are easy to open and close
  • Compatible with thermal cyclers accepting low-profile 0.2mL tubes
  • Ultra Thin Wall (UTW) low profile for fast PCR applications, compatible with the 24-well Piko Thermal Cycler and PikoReal Real-Time PCR System
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Color White
Description With attached optical flat caps
Product Type Tube
Certified free from human DNA, RNases, and DNases.