Learn More
Thomas Scientific 0.1 mL Split-Strip 8-Strip Tubes w/Individually Attached Flat Caps, Low-Profile, Clear, 120/pk
SDP
Supplier:  Thomas Scientific 1148A26
Encompass_Preferred
8 Strip Split-Strip with attached individual caps. Split-a-part design is easy to break allowing you to use only the tubes that you need. Certified free of detectable RNase DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. NC1809666