Learn More
Alkali Scientific 0.1mL PCR 8strip Tubes, Low Profile, Clear, No Caps, 125/pk, 1250/cs
SDP
Supplier:  Alkali Scientific PCN7055
Encompass_Preferred
Fast Cycler and Lightcycler compatible. Angularly attached caps on UltraFlux i and SnapStrip tubes prevent cap lids and hinges from interfering with each other, allowing for easier operation in an SBS format rack. Ultra-thin walls ensure efficient thermal transfer and maximum yield, yet are strong enough to prevent crushing. Clear tubes provide high clarity for visualizing contents. White tubes offer enhanced fluorescence reflection and the optically-clear caps, thanks to proprietary two-material manufacturing, ensure that the fluorescence signal is passed through uninhibited, creating the perfect environment for consistent and reliable qPCR performance.For use in pharmaceutical industry, food industry, and molecular biology. Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity. Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Matching caps are SKU PCN7056.
Catalog No. 50-238-8917
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.