Learn More
Thomas Scientific 0.2mL Rigid Strip 8-Strip Tubes w/Individually Attached Caps, Clear, 120/pack
SDP
Supplier:  Thomas Scientific 1148A28
Encompass_Preferred
8 Strip Rigid Strips with attached individual caps. Available in two sizes to fit virtually every thermal cycler or real-time instrument. 0.2mL for standard blocks and 0.1mL for low-profile or "fast" blocks.. Available in clear, assorted, or white. All caps are flat and optically-clear for realtime PCR assays. Rigid design is easy to work with minimizing the need to work with a PCR rack. Conveniently stacked in bags of 12 strips per bag (10 bags per unit) minimizes the risk of contamination. Certified free of detectable RNase DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. NC0926537