Learn More
Thomas Scientific 0.2mL Split-Strip 8-Strip Tubes w/Individually Attached Dome Cap, Clear (120/pk)
SDP
Supplier:  Thomas Scientific 1148A16
Encompass_Preferred
8 Quickstrip Clear, 0.2ml PCR Tubes w/ Individual Domed Caps, Certified RNase, DNase and Pyrogen free, 120 strips/unit
Catalog No. NC9909941